ContactUsLinkComponent
Unions for hplc
Unions for hplc
Catalog # MICS49302-03
Supplier:  MICROSOLV
undefined
Unions for hplc
Catalog # MICS49302-03
Supplier:  MICROSOLV
Specifications
  • Description:
    Union for HPLC, body only, Hole size: 0,254 mm
  • Material:
    PEEK
  • Pk:
    1
Specifications
About this item
The endure™ brand of carbon peek high pressure fittings are designed for extra durability. These fittings will endure higher temperatures, solvent exposure and over-tightening longer than standard peek fittings.
  • Low dead volume