Buffered peptone water, dehydrated culture media
Catalog # HIMEM1494I-500G
Anbieter: HIMEDIA
CAS-Nummer:
Technische Daten
- Pk:500 g
- Description:Buffered peptone water
- Seq:01
Technische Daten
Über diesen Artikel
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
Certifications: ISO 6579-2017 , ISO 6887 and ISO 21528-2017.