ContactUsLinkComponent
C.T. agar dehydrated culture media
C.T. agar dehydrated culture media
  HIMEM1349-500G
 :  HIMEDIA
 :  
C.T. agar dehydrated culture media
  HIMEM1349-500G
 :  HIMEDIA
 :  
  • Pk:
    500 g
  • Description:
    C.T. agar
For the cultivation of myxobacteria.
  • Microbiological culture media
  • 500 g poly bottles