ContactUsLinkComponent
Bird seed agar, dehydrated culture media
Bird seed agar, dehydrated culture media
Catalog # HIMEM675-500G
Supplier:  HIMEDIA
CAS Number:  
Bird seed agar, dehydrated culture media
Catalog # HIMEM675-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Bird seed agar (staib’s medium)
Specifications
About this item
For the selective isolation and differentiation of Cryptococcus neoformans from other Cryptococcus and other yeasts.
  • Microbiological culture media
  • Packaged in 500 g poly bottles