ContactUsLinkComponent
EMB agar dehydrated culture media
EMB agar dehydrated culture media
Catalog # HIMEM317-500G
Supplier:  HIMEDIA
CAS Number:  
EMB agar dehydrated culture media
Catalog # HIMEM317-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    EMB agar
Specifications
About this item
For the differential isolation of gram-negative Enteric bacilli from clinical and nonclinical specimens.
  • Microbiological culture media
  • 500 g poly bottles