Order Entry
Switzerland
ContactUsLinkComponent
 
Gifu anaerobic broth
  HIMEM1801-500G
 :  HIMEDIA
 :  
Gifu anaerobic broth
  HIMEM1801-500G
 :  HIMEDIA
 :  

 

  • Pk:
    500 g
  • Description:
    Gifu anaerobic broth (GAM)
  • Storage temperature:
    2 - 8 °C

 

 

Gifu Anaerobic Medium (GAM Broth) is recommended as a general culture medium for cultivation and isolation of anaerobic bacteria and to test their susceptibility to antibiotics other than sulpha drugs.

  • Light yellow to brownish yellow homogeneous free flowing powder
  • Forms amber coloured clear solution in tube
  • Reaction of 5,9% w/v aqueous solution at 25 °C, pH: 7,3 ±0,1

Composition of the medium is as follows: Peptic digest of animal tissue, papaic digest of soyabean meal, protease peptone, digested serum, yeast extract, beef extract, liver extract, dextrose, potassium dihydrogen phosphate, sodium chloride, starch, L-cysteine hydrochloride, sodium thioglycollate.

GAM broth is to be storted below 30 °C and the prepared medium at 2...8 °C.