Order Entry
Switzerland
ContactUsLinkComponent
Acetonitrile : Water (50:50) %, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Irritant
Flammable
Acetonitrile : Water (50:50) %, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC435-4
CAS Number:  
Irritant
Flammable
Acetonitrile : Water (50:50) %, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC435-4
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5186
  • Pk Info:
    4x 4 L

Specifications

About this item

 :
Flash Pt:3 °C
Storage Temperature:Ambient
EINECS:203-466-5
UN:1648
ADR:3,II
 :GHS07
 :GHS02
 :7732-18-5