Order Entry
Switzerland
ContactUsLinkComponent
Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Irritant
Health
Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Certificates
Irritant
Health
Dichloromethane ≥99.9% stabilised, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 20 ppm
  • Refractive index:
    1.4236 - 1.4246
  • Residue from evaporation:
    Max. 1 mg/l
  • Non-volatile matter:
    Max. 1 mg/l
  • Free acid (as HCl):
    Max. 1 ppm
  • Absorbance at 240 nm:
    Max. 0.1
  • Absorbance at 250 nm:
    Max. 0.01
  • UV- cutoff:
    Max. 233 nm
  • Chloride (Cl):
    Max. 10 ppm

Specification Test Results

Frequently Bought Together