ContactUsLinkComponent
Qualitative filter papers, Grade 598 ½, Whatman™
Qualitative filter papers, Grade 598 ½, Whatman™
Certificates
undefined
Qualitative filter papers, Grade 598 ½, Whatman™
Specifications
  • Catalog No:
  • 515-5211
  • 515-5204
  • 515-5207
  • 512-3635
  • Grade:
  • 598 ½
  • 598 ½
  • 598 ½
  • 598 ½
  • Weight:
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • 140 g/m²
  • Thickness:
  • 320 µm
  • 320 µm
  • 320 µm
  • 320 µm
  • Particle retention:
  • 10 µm
  • 10 µm
  • 10 µm
  • 10 µm
  • Filtration speed:
  • 50 s (Herzberg)
  • 50 s (Herzberg)
  • 50 s (Herzberg)
  • 50 s (Herzberg)