Order Entry
Switzerland
ContactUsLinkComponent
Tetrahydrofuran ≥99.9%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Flammable
Health
Irritant
Tetrahydrofuran ≥99.9%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Certificates
Flammable
Health
Irritant
Tetrahydrofuran ≥99.9%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Water (Karl Fischer):
    Max. 100 ppm
  • Refractive index:
    1.4065 - 1.4075
  • Peroxides (As H2O2) At Time Of Packaging:
    Max. 2 mg/l

Specification Test Results

Frequently Bought Together