Order Entry
Switzerland
ContactUsLinkComponent
Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Irritant
Health
Flammable
Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™
Certificates
Irritant
Health
Flammable
Pyridine ≥99.8%, BioSyn™ for biosynthesis, for applications requiring extremely low water content, Burdick & Jackson™

Specification Test Results

  • Amines by Ninhydrin Test:
    10 ppm max.
  • Free Acid:
    Passes test
  • GC Analysis:
    99.8 % min.
  • Refractive Index (20ºC):
    1.5085 - 1.5109
  • Residue:
    1 mg/L max.
  • Water by Karl Fischer Titration:
    50 ppm max.
  • Amines By Ninhydrin:
    Max. 10 ppm

Specification Test Results

Frequently Bought Together