n-Heptane ≥99.0% (by GC), Purum
51750-25L
:
- Pack type:Steel drum
- Pk:25 L
- Seq:5327
- Appearance (color):complying
- Appearance (form):complying
- IR spectrum:complying
- Assay (GC):Min. 99.0 %
- Density (D 20/4):0.682 - 0.686
- Refractive index (n 20/D):1.387 - 1.389
- Water (Karl Fischer):Max. 0.01 %
:
:100.20 g/mol
:98.2…98.4 °C (1,000 hPa)
:–90.5 °C
:0.69 g/cm3 (15 °C)
:–4 °C
:1206
:3,II
:142-82-5
:H₃C(CH₂)₅CH₃
:MFCD00009544