Order Entry
Switzerland
ContactUsLinkComponent
LC-SDA (NHS-LC-Diazirine) (succinimidyl 6-(4,4′-azipentanamido)hexanoate), Pierce™
LC-SDA (NHS-LC-Diazirine) (succinimidyl 6-(4,4′-azipentanamido)hexanoate), Pierce™
Catalog # PIER26168
CAS Number:  
undefined
LC-SDA (NHS-LC-Diazirine) (succinimidyl 6-(4,4′-azipentanamido)hexanoate), Pierce™
Catalog # PIER26168
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    NHS Ester, Diazirine
  • Spacer:
    Long
  • Form:
    Solid
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO
  • Spacer Arm:
    12.5 Å

Specification Test Results

About this item

 :
 :338.36 g/mol
Storage Temperature:Refrigerator
Formula:C₁₅H₂₂N₄O₅