Pierce™ Deuterium-labelled Cross Linkers
Supplier: Thermo Fisher Scientific
Certificates
About this item
Cross linkers are used for the identification of near-neighbor protein relationships and ligand-receptor interactions.
- Deuterium-labelled cross linkers
Deuterium-labelled cross linkers are specially manufactured for crosslinking protein structure: Function analysis by mass spectrometry.
Frequently Bought Together