Erythromycin Ethylsuccinate, Reference Standard
Erythromycin Ethylsuccinate, USP reference standard, Vial, 200 mg
1245008.
: USP
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Pack type:Vial
- Pk:200 mg
- Standards:USP reference standard
- Description:Erythromycin Ethylsuccinate
- Cat. No.:1245008.
- Formula:C₄₃H₇₅NO₁₆
- Description:Erythromycin Ethylsuccinate
- CAS No.:1264-62-6



