Order Entry
Switzerland
ContactUsLinkComponent
 
Citalopram Citalopram Related Compounds, Reference Standard
 :  USP
 
Citalopram Citalopram Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    1188264-72-3
  • Formula:
    C₁₉H₂₀BrFN₂O
  • Description:
    Citalopram related compound D; 1-(4-Fluorophenyl)-1-(3-methylaminopropyl) -1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide

 

Frequently Bought Together