

Benzoic acid, Fluka™
Catalog # 32035-6X500G
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:3.000 g
- Seq:3810
Specifications
Specification Test Results
- suitability (for KF-titration):test passed
- assay of water:Max. 0.2 %
- number of samples:Min. 2
Specification Test Results
About this item
:
:122.12 g/mol
Boiling Pt:249-250 °C
Boiling Pt:249…250 °C
Melting Pt:121-123 °C
Melting Pt:121…123 °C
Density:1.27…1.32 g/cm³ (20 °C)
Flash Pt:121 °C
EINECS:200-618-2
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:65-85-0
Formula:C₇H₆O₂
MDL Number:MFCD00002398