Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Polyoxyl Lauryl Ether, Reference Standards

Polyoxyl Lauryl Ether, Reference Standards

Supplier:   USP
Polyoxyl Lauryl Ether, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1372424.
  • 1372468.
  • 1372435.
  • Description:
  • Polyoxyl 4 Lauryl Ether
  • Polyoxyl 23 Lauryl Ether
  • Polyoxyl 9 Lauryl Ether
  • Synonym(s):
  • Polyethylene glycol monolauryl ether (n=9)
  • CAS No.:
  • 5274-68-0
  • 9002-92-0
  • 9002-92-0
  • UN No.:
  • 3082
  • 3082
  • 3082

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1372424.
    Polyoxyl 4 Lauryl Ether
    Polyoxyl 4 Lauryl Ether
    USP reference standard
    Ampule
    1 mL
    Log in for availability
    Log in for availability
    1 * 1 mL
    1 * 1 mL - 210.00 SFr.
    210.00 SFr.
    1 * 1 mL
       210.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1372424.
    Description
    Polyoxyl 4 Lauryl Ether
    Synonym(s)
    CAS No.
    5274-68-0
    UN No.
    3082
  • 1372468.
    Polyoxyl 23 Lauryl Ether
    Polyoxyl 23 Lauryl Ether
    USP reference standard
    Vial
    0,5 g
    Log in for availability
    Log in for availability
    1 * 0,5 g
    1 * 0,5 g - 426.00 SFr.
    426.00 SFr.
    1 * 0,5 g
       426.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1372468.
    Description
    Polyoxyl 23 Lauryl Ether
    Synonym(s)
    CAS No.
    9002-92-0
    UN No.
    3082
  • 1372435.
    Polyoxyl 9 Lauryl Ether
    Polyoxyl 9 Lauryl Ether
    USP reference standard
    Vial
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 325.00 SFr.
    325.00 SFr.
    1 * 1 g
       325.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1372435.
    Description
    Polyoxyl 9 Lauryl Ether
    Synonym(s)
    Polyethylene glycol monolauryl ether (n=9)
    CAS No.
    9002-92-0
    UN No.
    3082
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.