Polyoxyl Stearyl Ether, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
4 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Certificates
Search by Lot NumberSpecifications for Product Family
Specifications
Specifications
- Catalog No:
- 1372606.
- 1547346.
- 1547368.
- 1547801.
- Description:
- Polyoxyl 10 Stearyl Ether
- Polyoxyl 2 Stearyl Ether
- Polyoxyl 20 Stearyl Ether
- Polyoxyl 20 Cetostearyl Ether
- CAS No.:
- 9005-00-9
- 9005-00-9
- 9005-00-9
- 68439-49-6
- Formula:
- C₂₀H₄₂O₂
- C₂₀H₄₂O₂
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1372606.Polyoxyl 10 Stearyl EtherSpecifications:
More
Specifications
Cat. No.1372606.DescriptionPolyoxyl 10 Stearyl EtherCAS No.9005-00-9Formula - 1547346.Polyoxyl 2 Stearyl EtherSpecifications:
More
Specifications
Cat. No.1547346.DescriptionPolyoxyl 2 Stearyl EtherCAS No.9005-00-9FormulaC₂₀H₄₂O₂ - 1547368.Polyoxyl 20 Stearyl EtherSpecifications:
More
Specifications
Cat. No.1547368.DescriptionPolyoxyl 20 Stearyl EtherCAS No.9005-00-9FormulaC₂₀H₄₂O₂ - 1547801.Polyoxyl 20 Cetostearyl EtherPolyoxyl 20 Cetostearyl EtherUSP reference standardVial100 mgLog in for availability1 * 100 mgSFr. 485.00Specifications:
More
Specifications
Cat. No.1547801.DescriptionPolyoxyl 20 Cetostearyl EtherCAS No.68439-49-6Formula
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



