Order Entry
Switzerland
ContactUsLinkComponent
 
Timolol Maleate, Reference Standard
  1667406.
 :  USP
 :  
Timolol Maleate, Reference Standard
  1667406.
 :  USP
 :  

 

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • CAS No.:
    26921-17-5
  • Description:
    Timolol Maleate
  • Seq:
    07
  • Environmentally Preferable:
  • Formula:
    C₁₇H₂₈N₄O₇S
  • Description:
    Timolol Maleate

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard