Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Pancuronium Bromide, Reference Standards

Pancuronium Bromide, Reference Standards

Supplier:   USP
Pancuronium Bromide, Reference Standards
Supplier:  USP
Certificates

4 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1494217.
  • 1494228.
  • 1494239.
  • 1494240.
  • Description:
  • Pancuronium Bromide
  • Pancuronium Bromide Related Compound A
  • Pancuronium Bromide Related Compound B
  • Pancuronium Bromide Related Compound C
  • Synonym(s):
  • 1,1'-(3α,17β-dihydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
  • 1,1'-(17β-Acetoxy-3α-hydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
  • 1,1'-(3α-acetoxy-17β-hydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
  • CAS No.:
  • 15500-66-0
  • 15500-65-9
  • 41261-71-6
  • 27115-86-2
  • UN No.:
  • 2811
  • 2811
  • 2811
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1494217.
    Pancuronium Bromide
    Pancuronium Bromide
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 711.00 SFr.
    711.00 SFr.
    1 * 500 mg
       711.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1494217.
    Description
    Pancuronium Bromide
    Synonym(s)
    CAS No.
    15500-66-0
    UN No.
    2811
  • 1494228.
    Pancuronium Bromide Related Compound A
    Pancuronium Bromide Related Compound A
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 813.00 SFr.
    813.00 SFr.
    1 * 15 mg
       813.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1494228.
    Description
    Pancuronium Bromide Related Compound A
    Synonym(s)
    1,1'-(3α,17β-dihydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
    CAS No.
    15500-65-9
    UN No.
    2811
  • 1494239.
    Pancuronium Bromide Related Compound B
    Pancuronium Bromide Related Compound B
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 360.00 SFr.
    1 360.00 SFr.
    1 * 15 mg
       1 360.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1494239.
    Description
    Pancuronium Bromide Related Compound B
    Synonym(s)
    1,1'-(17β-Acetoxy-3α-hydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
    CAS No.
    41261-71-6
    UN No.
    2811
  • 1494240.
    Pancuronium Bromide Related Compound C
    Pancuronium Bromide Related Compound C
    USP reference standard
    Vial
    15 mg
    Log in for availability
    Log in for availability
    1 * 15 mg
    1 * 15 mg - 1 270.00 SFr.
    1 270.00 SFr.
    1 * 15 mg
       1 270.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1494240.
    Description
    Pancuronium Bromide Related Compound C
    Synonym(s)
    1,1'-(3α-acetoxy-17β-hydroxy-5α-androstan-2β,16β-ylene) bis(1-methylpiperidinium) dibromide
    CAS No.
    27115-86-2
    UN No.
    2811