Iohexol, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1344600.
- 1344622.
- 1344644.
- 1344666.
- Description:
- Iohexol
- Iohexol Related Compound A
- Iohexol Related Compound B
- Iohexol Related Compound C
- Synonym(s):
- 5-(acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
- 5-amino-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
- N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamide
- CAS No.:
- 66108-95-0
- 31127-80-7
- 76801-93-9
Product Family Options
- DescriptionStandardsPack typePk
- IohexolUSP reference standardVial200 mg1 * 200 mgSFr. 530.00
Specifications
Cat. No.1344600.DescriptionIohexolSynonym(s)CAS No.66108-95-0 - Iohexol Related Compound AUSP reference standardVial100 mg1 * 100 mgSFr. 1’310.00
Specifications
Cat. No.1344622.DescriptionIohexol Related Compound ASynonym(s)5-(acetylamino)-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamideCAS No.31127-80-7 - Iohexol Related Compound BUSP reference standardVial50 mg1 * 50 mgSFr. 1’410.00
Specifications
Cat. No.1344644.DescriptionIohexol Related Compound BSynonym(s)5-amino-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamideCAS No.76801-93-9 - Iohexol Related Compound CUSP reference standardVial50 mg1 * 50 mgSFr. 1’370.00
Specifications
Cat. No.1344666.DescriptionIohexol Related Compound CSynonym(s)N,N'-bis(2,3-dihydroxypropyl)-5-nitro-1,3-benzenedicarboxamideCAS No.



