Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Iodixanol, Reference Standards

Iodixanol, Reference Standards

 :   USP
Iodixanol, Reference Standards
 :  USP
 
Iodixanol, Reference Standards

 

 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

 

  • Catalog No:
  • 1343517.
  • 1343540.
  • 1343550.
  • 1343561.
  • Description:
  • Iodixanol
  • Iodixanol Related Compound C
  • Iodixanol Related Compound D
  • Iodixanol Related Compound E
  • Synonym(s):
  • 5-[Acetyl[3-[[3,5-bis[[(2,3-dihydroxypropyl)amino]carbonyl]-2,4,6-triiodophenyl]amino]-2-hydroxypropyl]amino]-N,N'-bis-(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
  • 5-[Acetyl(2-hydroxy-3-methoxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
  • 5-{N-[3-(N-{3-carbamoyl-5-[(2,3-dihydroxypropyl)carbamoyl]-2,4,6-triiodophenyl}acetamido)-2-hydroxypropyl]acetamido}-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
  • CAS No.:
  • 92339-11-2
  • 171897-74-8
  • 89797-00-2
  • 255376-57-9

 

Product Family Options

 

  • Description
    Standards
    Pack type
    Pk
     
     
  • Iodixanol
    USP reference standard
    Vial
    200 mg
       
       
    1 * 200 mg
    1 * 200 mg - 436.00 SFr.
    436.00 SFr.
    1 * 200 mg
       436.00 SFr.

     
     
    Specifications
    Cat. No.
    1343517.
    Description
    Iodixanol
    Synonym(s)
    CAS No.
    92339-11-2
  • Iodixanol Related Compound C
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 420.00 SFr.
    1 420.00 SFr.
    1 * 25 mg
       1 420.00 SFr.

     
     
    Specifications
    Cat. No.
    1343540.
    Description
    Iodixanol Related Compound C
    Synonym(s)
    5-[Acetyl[3-[[3,5-bis[[(2,3-dihydroxypropyl)amino]carbonyl]-2,4,6-triiodophenyl]amino]-2-hydroxypropyl]amino]-N,N'-bis-(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
    CAS No.
    171897-74-8
  • Iodixanol Related Compound D
    USP reference standard
    Vial
    50 mg
       
       
    1 * 50 mg
    1 * 50 mg - 1 480.00 SFr.
    1 480.00 SFr.
    1 * 50 mg
       1 480.00 SFr.

     
     
    Specifications
    Cat. No.
    1343550.
    Description
    Iodixanol Related Compound D
    Synonym(s)
    5-[Acetyl(2-hydroxy-3-methoxypropyl)amino]-N,N'-bis(2,3-dihydroxypropyl)-2,4,6-triiodo-1,3-benzenedicarboxamide
    CAS No.
    89797-00-2
  • Iodixanol Related Compound E
    USP reference standard
    Vial
    25 mg
       
       
    1 * 25 mg
    1 * 25 mg - 1 480.00 SFr.
    1 480.00 SFr.
    1 * 25 mg
       1 480.00 SFr.

     
     
    Specifications
    Cat. No.
    1343561.
    Description
    Iodixanol Related Compound E
    Synonym(s)
    5-{N-[3-(N-{3-carbamoyl-5-[(2,3-dihydroxypropyl)carbamoyl]-2,4,6-triiodophenyl}acetamido)-2-hydroxypropyl]acetamido}-N1,N3-bis(2,3-dihydroxypropyl)-2,4,6-triiodoisophthalamide
    CAS No.
    255376-57-9