Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Gemfibrozil, Reference Standards

Gemfibrozil, Reference Standards

Supplier:   USP
Gemfibrozil, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1288500.
  • 1288510.
  • Description:
  • Gemfibrozil
  • Gemfibrozil Related Compound A
  • Synonym(s):
  • (E,Z)-2,2-dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid
  • CAS No.:
  • 25812-30-0
  • 500904-61-0
  • Formula:
  • C₁₈H₂₆O₃

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1288500.
    Gemfibrozil
    Gemfibrozil
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 403.00 SFr.
    403.00 SFr.
    1 * 200 mg
       403.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1288500.
    Description
    Gemfibrozil
    Synonym(s)
    CAS No.
    25812-30-0
    Formula
  • 1288510.
    Gemfibrozil Related Compound A
    Gemfibrozil Related Compound A
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - 1 400.00 SFr.
    1 400.00 SFr.
    1 * 20 mg
       1 400.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1288510.
    Description
    Gemfibrozil Related Compound A
    Synonym(s)
    (E,Z)-2,2-dimethyl-5-[2,5-dimethyl-4-(propene-1-yl)phenoxy]valeric acid
    CAS No.
    500904-61-0
    Formula
    C₁₈H₂₆O₃