Ethacrynic Acid, Reference Standards
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Catalog No:
- 1256004.
- 1256015.
- 1256037.
- Description:
- Ethacrynic Acid
- Ethacrynic Acid Related Compound A
- Ethacrynic Acid Related Compound C
- Synonym(s):
- (4-Butyryl-2,3-dichlorophenoxy)acetic acid
- {4-[2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy}acetic acid
- CAS No.:
- 58-54-8
- 1217-67-0
- 25355-92-4
Product Family Options
- DescriptionStandardsPack typePk
- Ethacrynic AcidUSP reference standardVial200 mg1 * 200 mgSFr. 439.00
Specifications
Cat. No.1256004.DescriptionEthacrynic AcidSynonym(s)CAS No.58-54-8 - Ethacrynic Acid Related Compound AUSP reference standardVial25 mg1 * 25 mgSFr. 1’440.00
Specifications
Cat. No.1256015.DescriptionEthacrynic Acid Related Compound ASynonym(s)(4-Butyryl-2,3-dichlorophenoxy)acetic acidCAS No.1217-67-0 - Ethacrynic Acid Related Compound CUSP reference standardVial25 mg1 * 25 mgSFr. 1’440.00
Specifications
Cat. No.1256037.DescriptionEthacrynic Acid Related Compound CSynonym(s){4-[2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy}acetic acidCAS No.25355-92-4



