Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Eletriptan Related Compounds, Reference Standard

Eletriptan Related Compounds, Reference Standard

Supplier:   USP
Eletriptan Related Compounds, Reference Standard
Supplier:  USP
Certificates
Eletriptan Related Compounds, Reference Standard

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1234453.
  • 1234475.
  • 1234522.
  • Description:
  • Eletriptan Hydrobromide
  • Eletriptan Related Compound B
  • Eletriptan S-Enantiomer
  • Synonym(s):
  • 3-[[(R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]indole monohydrobromide monohydrate
  • (R)-5-Bromo-3-[(1-methylpyrrolidin-2-yl)methyl]-1H-indole
  • (S)-3-[(1-methylpyrrolidin-2-yl)methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole, monohydrobromide
  • CAS No.:
  • 273211-28-2
  • 143322-57-0
  • UN No.:
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1234453.
    Eletriptan Hydrobromide
    Eletriptan Hydrobromide
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 770.00
    SFr. 770.00
    1 * 200 mg
       SFr. 770.00

    Specifications:
    More
    Specifications
    Cat. No.
    1234453.
    Description
    Eletriptan Hydrobromide
    Synonym(s)
    3-[[(R)-1-Methyl-2-pyrrolidinyl]methyl]-5-[2-(phenylsulfonyl)ethyl]indole monohydrobromide monohydrate
    CAS No.
    273211-28-2
    UN No.
  • 1234475.
    Eletriptan Related Compound B
    Eletriptan Related Compound B
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - SFr. 805.00
    SFr. 805.00
    1 * 20 mg
       SFr. 805.00

    Specifications:
    More
    Specifications
    Cat. No.
    1234475.
    Description
    Eletriptan Related Compound B
    Synonym(s)
    (R)-5-Bromo-3-[(1-methylpyrrolidin-2-yl)methyl]-1H-indole
    CAS No.
    143322-57-0
    UN No.
    3077
  • 1234522.
    Eletriptan S-Enantiomer
    Eletriptan S-Enantiomer
    USP reference standard
    Vial
    20 mg
    Log in for availability
    Log in for availability
    1 * 20 mg
    1 * 20 mg - SFr. 1’360.00
    SFr. 1’360.00
    1 * 20 mg
       SFr. 1’360.00

    Specifications:
    More
    Specifications
    Cat. No.
    1234522.
    Description
    Eletriptan S-Enantiomer
    Synonym(s)
    (S)-3-[(1-methylpyrrolidin-2-yl)methyl]-5-[2-(phenylsulfonyl)ethyl]-1H-indole, monohydrobromide
    CAS No.
    UN No.