Order Entry
Switzerland
ContactUsLinkComponent
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], Pierce™
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], Pierce™
Catalog # PIER21565
CAS Number:  
undefined
EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)], Pierce™
Catalog # PIER21565
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    NHS Ester
  • Spacer:
    Cleavable, Long
  • Form:
    Powder
  • Quantity:
    1 g
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO

Specification Test Results

About this item

 :
 :456.36 g/mol
Storage Temperature:Refrigerator
CAS Number:70539-42-3
Formula:C₁₈H₂₀N₂O₁₂
MDL Number:MFCD00049057