EGS [Ethylene glycolbis(succinic acid-N-hydroxysuccinimide ester)]
Catalog # BC09
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Spacer Arm (Å):16.1
- Reactive Toward:Primary Amines
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:Hydroxylamine
Specification Test Results
About this item
:
:456.36 g/mol
CAS Number:70539-42-3
Formula:C₁₈H₂₀N₂O₁₂
MDL Number:MFCD00049057