ContactUsLinkComponent
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
  BC20
 :  
undefined
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
  BC20
 :  
  • Pk:
    100 mg
  • Seq:
    2440
  • Spacer Arm (Å):
    11.6
  • Reactive Toward:
    Primary Amine + Sulfhydryl
  • Membrane Permeable:
    Yes
  • Water Soluble:
    No
  • Cleavable/ Reversible:
    No
 :
 :334.33 g/mol
 :64987-85-5
 :C₁₆H₁₈N₂O₆
 :MFCD00009634