GMBS (N-Succinimidyl 4-maleimidobutyrate)
Catalog # BC13
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Spacer Arm (Å):6.8
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:Yes
- Water Soluble:No
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:280.24 g/mol
CAS Number:80307-12-6
Formula:C₁₂H₁₂N₂O₆
MDL Number:MFCD00036817