Order Entry
Switzerland
ContactUsLinkComponent
 
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), OneQuant™
Catalog # 786-082
Supplier:  G-Biosciences
CAS Number:  
undefined
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), OneQuant™
Catalog # 786-082
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    8
  • Seq:
    0008
  • Pk Info:
    8x5 mg

Specifications

Specification Test Results

  • Spacer Arm (Å):
    11.6
  • Reactive Toward:
    Primary Amine + Sulfhydryl
  • Membrane Permeable:
    No
  • Water Soluble:
    Yes
  • Cleavable/ Reversible:
    No

Specification Test Results

About this item

 :
 :436.37 g/mol
 :GHS07
CAS Number:92921-24-9
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978