Order Entry
Switzerland
ContactUsLinkComponent
 
Swinging-bucket rotor, SW 55 Ti, Optima
Catalog # 521-3730
Supplier:  Beckman Coulter
undefined
Swinging-bucket rotor, SW 55 Ti, Optima
Catalog # 521-3730
Supplier:  Beckman Coulter

Specifications

  • Description:
    SW 55 Ti swinging bucket rotor set
  • Pk:
    1
  • For:
    Swinging bucket (ultracentrifugation)
  • Max. RPM:
    55000
  • No. of tubes accepted:
    6×5 ml / 13×51 mm (6×5 ml / 1×2")
  • Radius:
    108,5 mm

Specifications

About this item