0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Centrifuges
Rotors
Swinging bucket rotor Assembly, SW 40 Ti, Optima
Swinging bucket rotor Assembly, SW 40 Ti, Optima
Swinging bucket rotor Assembly, SW 40 Ti, Optima
Catalog # 521-3569
Supplier:
Beckman Coulter
Expand image
Swinging bucket rotor Assembly, SW 40 Ti, Optima
Catalog # 521-3569
Supplier:
Beckman Coulter
Specifications
Description:
SW 40 Ti swinging bucket rotor assembly
Pk:
1
For:
Swinging bucket (ultracentrifugation)
Max. RPM:
40000
Specifications
About this item
Product Details
Product Details