SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Centrifuges
Rotors
Zonal Rotor without core, Titanium, Ti-15, Optima
Zonal Rotor without core, Titanium, Ti-15, Optima
521-4069
:
Beckman Coulter
Zonal Rotor without core, Titanium, Ti-15, Optima
521-4069
:
Beckman Coulter
Description:
Ti-15 zonal rotor without core, titanium. Major applications: Rate-zonal centrifugation of subcellular particles
Pk:
1
For:
Speciality (ultracentrifugation)
Max. RPM:
32000