ContactUsLinkComponent
Noryl rotor, Core- Ti-15, Optima
Noryl rotor, Core- Ti-15, Optima
Catalog # 521-3585
Supplier:  Beckman Coulter
Noryl rotor, Core- Ti-15, Optima
Catalog # 521-3585
Supplier:  Beckman Coulter
Specifications
  • Description:
    Noryl rotor core- Ti-15 rotor, O-ring, top of rotor core (815472), O-ring for seal fitting (011167) O-ring, bottom of rotor core cone (812715) Cone (332682) O-ring, interior of Rotor Core (011519), O-ring, interior of cap, seal fitting (332691) and O-ring, visible, on cap (012780)
  • Pk:
    1
  • For:
    Rotor core, Noryl, For use in ultracentrifuge rotors