Order Entry
Switzerland
ContactUsLinkComponent
 
Rotor assembly with labware kit, Avanti JS-5
Catalog # 521-4047
Supplier:  Beckman Coulter
undefined
Rotor assembly with labware kit, Avanti JS-5
Catalog # 521-4047
Supplier:  Beckman Coulter

Specifications

  • Capacity:
    Nominal capacity
    9 L
  • Description:
    JS-5.0 swinging bucket (high performance) rotor package. For use in Avanti J-HC centrifuges. Package includes rotor body and lid with four removable buckets and labware kit: Red ccover gaskets, for use without liners (369257 set of 4); green cover gaskets with liners (369261 set of 4); air vent filters (368148 set of 60); spatulas (367891 set of 2), 2× cup racks 4 J-wide cups and cup covers with plugs
  • Pk:
    1
  • For:
    Major applications: Separating bacterial, yeast, and tissue homogenates; harvesting cultures
  • Max. RPM:
    5000
  • Radius:
    267 mm

Specifications

About this item