Order Entry
Switzerland
ContactUsLinkComponent
Swinging-bucket rotor and buckets, Avanti JS5.3
Swinging-bucket rotor and buckets, Avanti JS5.3
Catalog # 521-4053
Supplier:  Beckman Coulter
undefined
Swinging-bucket rotor and buckets, Avanti JS5.3
Catalog # 521-4053
Supplier:  Beckman Coulter

Specifications

  • Capacity:
    Nominal capacity
    2000 ml
  • Description:
    JS-5.3 AllSpin swinging bucket (high performance) rotor and buckets. For use in Avanti J-E and J-26 XP Series centrifuges only. Includes tie-down knob.
  • Pk:
    1
  • For:
    Four place rotor with anodised aluminium buckets designed for use with adapters to accommodate a wide range of tubes and bottles. Rotor buckets are interchangeable with microplate carriers. Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and separating serum from whole blood
  • Max. RPM:
    5300
  • Radius:
    500 ml conical: 218,4 mm, Deep-well Plates: 194,8 mm

Specifications

About this item