Order Entry
Switzerland
ContactUsLinkComponent
Rotor, Avanti JS-4.3
Rotor, Avanti JS-4.3
Catalog # 521-3989
Supplier:  Beckman Coulter
undefined
Rotor, Avanti JS-4.3
Catalog # 521-3989
Supplier:  Beckman Coulter

Specifications

  • Capacity:
    Nominal capacity
    3 L
  • Description:
    JS-4.3 swinging bucket (high performance) stainless steel rotor and buckets. For use in Avanti J-26 XP Series and J-HC centrifuges.For use in Avanti J-26 XP Series and J-HC centrifuges
  • Pk:
    1
  • For:
    Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and separating serum from whole blood
  • Max. RPM:
    4300
  • No. of tubes accepted:
    4×750 ml/96×130 mm (4×750 ml/4×5")
  • Radius:
    204 mm

Specifications

About this item

Four-place rotor with anodised aluminium buckets designed for use with modular disk adapters to accommodate a variety of tubes and other adapters and blood bag cups. Can be used with Beckman Coulter aerosolve canisters which are designed to provide added protection from aerosols. Rotor buckets are interchangeable with microplate carriers.