Rotor assembly, Avanti JS-4.2
Catalog # 521-3705
Supplier: Beckman Coulter
Specifications
- Capacity:Nominal capacity
6 L - Description:JS-4.2 swinging bucket (high performance) aluminium rotor and buckets. For use in Avanti J-HC and J6 Series centrifuges only
- Pk:1
- For:Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and separating serum from whole blood
- Max. RPM:4200
- Radius:254 mm
Specifications
About this item
Windshielded, six-place rotor with aluminium rotor yoke and removable aluminium swinging buckets. Buckets are interchangeable with microplate carriers (358682) for spinning microtitre plates and minitube racks. Holds up to six litres, six blood bags, or six microtitre plate carriers. Accepts all multi-disc adapters. A kit is available to convert this rotor to a JS-4.2SM (356501)