Order Entry
Switzerland
ContactUsLinkComponent
Rotor assembly, Avanti JS-4.0
Rotor assembly, Avanti JS-4.0
Catalog # 521-3706
Supplier:  Beckman Coulter
Rotor assembly, Avanti JS-4.0
Catalog # 521-3706
Supplier:  Beckman Coulter

Specifications

  • Capacity:
    Nominal capacity
    4 L
  • Description:
    JS-4.0 swinging bucket (high performance) aluminium rotor and buckets. For use in Avanti J-26 XP Series, J-26S Series and J-6 Series centrifuges
  • Pk:
    1
  • For:
    Major applications: Rapid sedimentation of protein precipitates, large particles, cells, and cell debris. It can be used for binding studies and separating serum from whole blood
  • Max. RPM:
    4000
  • Radius:
    226 mm

Specifications

About this item

Unshielded, four-place rotor with aluminium rotor yoke and removable aluminium swinging buckets. Buckets are interchangeable with microplate carriers for spinning microtitre plates and minitube racks. Holds up to four litres, four blood bags or four microtitre plate carriers. Accepts all multi-disc adapters except 339119. Note: requires tie-down kit (367045) for use with Avanti J-26 XP Series centrifuges.

Caution: NOTE: Requires tie-down kit (367045) for use with Avanti J-26 XP Series centrifuges.