ContactUsLinkComponent
Fixed-angle aluminium rotor, Avanti JA-18.1
Fixed-angle aluminium rotor, Avanti JA-18.1
Supplier:  Beckman Coulter
Certificates
undefined
Fixed-angle aluminium rotor, Avanti JA-18.1
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-4197
  • 521-3816
  • Capacity:
  • 800 ml
  • 43,2 ml
  • No. of tubes accepted:
  • 16×50 ml conical
  • 24×1,8 ml microcentrifuge tubes
  • Radius:
  • 160 mm
  • 112 mm (45° angle), 116 mm (25° angle)
  • Max. RPM:
  • 14000
  • 18000