ContactUsLinkComponent
Swinging-bucket titanium rotor package/assembly, TLS-55
Swinging-bucket titanium rotor package/assembly, TLS-55
Supplier:  Beckman Coulter
Certificates
undefined
Swinging-bucket titanium rotor package/assembly, TLS-55
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-3802
  • 521-3804
  • Capacity:
  • 8,8 ml
  • 8,8 ml
  • No. of tubes accepted:
  • 4×2,2ml / 11×34 mm
  • 4×2,2ml / 11×34 mm
  • Radius:
  • 76,5 mm
  • 76,5 mm
  • Max. RPM:
  • 55000
  • 55000