Masterflex® Male Luer Plug Fitting, Avantor®
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297
Easily transition to a luer fitting.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances for increased reliability and process purity. All female ports may be used with either male slip or male luer lock connections. Utilizing a wide range of fitting materials, we can support applications requiring specific chemical compatibility.
CrystalVu:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.1520
Medical Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 178.3297
Natural Nylon:
Sterilization: Ethylene Oxide, Autoclave
Regulatory Approvals: FDA 21 CFR 177.1500
Natural Kynar:
Sterilization: Gamma irradiation, Ethylene Oxide, Autoclave
Regulatory Approvals: USP Class VI, FDA 21 CFR 177.2510
Polypropylene:
Sterilization: Gamma irradiation
Regulatory Approvals: FDA 21 CFR 178.3297
- Connection type (port 1):
- Male Luer Plug
- Male Luer Plug
- Male Luer Plug
- Male Luer Plug
- Male Luer Plug
- Male Luer Plug
- No. of ports:
- 1
- 1
- 1
- 1
- 1
- 1
- Regulatory approvals:
- FDA;USP
- FDA
- FDA;USP
- FDA
- FDA;USP
- FDA;USP
Product Family Options
- DescriptionConnection TypeFitting materialFlow pattern
- Male Luer plug in crystalVu cleanroom manufactured; 10 packMale Luer PlugCrystalVuStraight1 * 10 STSFr. 9.60
Specifications
Cat. No.MFLX40319-00Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA;USP - Male Luer plug in medical nylon cleanroom manufactured; 10 packMale Luer PlugMedical NylonStraight1 * 10 STSFr. 9.60
Specifications
Cat. No.MFLX40319-01Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA - Male Luer plug in natural PVDF cleanroom manufactured; 10 packMale Luer PlugPVDFStraight1 * 10 STSFr. 13.50
Specifications
Cat. No.MFLX40319-02Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA;USP - Male Luer plug in natural nylon cleanroom manufactured; 10 packMale Luer PlugNylonStraight1 * 10 STSFr. 9.60
Specifications
Cat. No.MFLX40319-03Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA - Male Luer plug in polypropylene cleanroom manufactured; 10 packMale Luer PlugPolypropyleneStraight1 * 10 STSFr. 14.00
Specifications
Cat. No.MFLX40319-04Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA;USP - Luer male with stopper plug in adf polypropylene cleanroom manufactured; 10 packMale Luer PlugPolypropyleneStraight1 * 10 STSFr. 38.00
Specifications
Cat. No.MFLX40319-05Connection type (port 1)Male Luer PlugNo. of ports1Regulatory approvalsFDA;USP