ContactUsLinkComponent
Masterflex® 3/4" Sanitary Clamp Low Flow Male Quick-Disconnect Fittings, Avantor®
Masterflex® 3/4" Sanitary Clamp Low Flow Male Quick-Disconnect Fittings, Avantor®
Certificates
undefined
Masterflex® 3/4" Sanitary Clamp Low Flow Male Quick-Disconnect Fittings, Avantor®
Specifications
  • O-Ring material:
  • Silicone
  • Viton
  • Silicone
  • Silicone
  • Viton
  • Viton
  • Viton
  • Silicone
  • Sterilisation:
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Gamma irradiation, EtO, autoclave
  • Valve spring material:
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Stainless Steel
  • Regulatory approvals:
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP
  • FDA;USP