Orders LinkContactUsLinkComponent
Tablet counting tray
Tablet counting tray
Supplier:   Sampling Systems
Tablet counting tray
Supplier:  Sampling Systems
Certificates
Tablet counting tray
Tablet counting tray
2 Options Available Below
Product Details & Documents
Makes easy work of counting round tablets.
  • Easy to use counting
  • Suitable for all sizes of round tablets
  • useful for both right-handed and left-handed operators
  • Satin finish
Counting round tablets has never been easier with the Sampling Systems tablet counting triangle. When you have finished counting, the intuitive desing means the tablets can be poured directly into the bottle.
Documents
Product Family Options
Product Information
  • Height
    Length
    Availability
    Price
  • 14 mm
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 137.00
    SFr. 137.00
    1 * 1 ST
       SFr. 137.00
  • 12 mm
    275 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 155.00
    SFr. 155.00
    1 * 1 ST
       SFr. 155.00