Order Entry
Switzerland
ContactUsLinkComponent
 
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Premium Grade, Pierce™
 
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt), Premium Grade, Pierce™

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Heterobifunctional
  • Reactive Moiety:
    Carbodiimide
  • Spacer:
    Short
  • Form:
    Powder
  • Quantity:
    500 mg /10 g
  • Type:
    Crosslinker
  • Solubility:
    Water

 

Frequently Bought Together