Order Entry
Switzerland
ContactUsLinkComponent
 
VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM
Catalog # 91132.001
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM
Catalog # 91132.001
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    100 ug/ml Carbendazim in methanol
  • Pack type:
    Glass ampoule
  • Pk:
    1 mL
  • CAS No.:
    10605-21-7
  • Description:
    LC Multiresidue Pesticide Standard (Mix 10)
  • UN No.:
    1992

Specifications

About this item

A standard, typically used in food testing, Carbendazim [CAS:10605-21-7] 100 ug/ml in Methanol

All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.

Certifications: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Caution: Storage temperature: Freezer (–20 °C)