Order Entry
Switzerland
ContactUsLinkComponent
VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 9) - 8 components, CRM
VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 9) - 8 components, CRM
Catalog # 91121.001
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® PESTINORM®, GC Multiresidue Pesticide Standard (Mix 9) - 8 components, CRM
Catalog # 91121.001
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    100 ug/ml each component in toluene
  • Pack type:
    Glass ampoule
  • Pk:
    1 mL
  • Description:
    GC Multiresidue Pesticide Standard (Mix 9)
  • UN No.:
    1294

Specifications

About this item

A standard mixture, typically used for food testing, containing the following components: 100 ug/ml each of Disulfoton [CAS:298-04-4] ; Fonofos [CAS:944-22-9] ; Parathion-methyl [CAS:298-00-0] ; Mevinphos [CAS:7786-34-7] ; Parathion [CAS:56-38-2] ; Phorate [CAS:298-02-2] ; Piperonyl butoxide [CAS:51-03-6] ; Triazophos [CAS:24017-47-8] in Toluene

All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.

Certifications: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Caution: Storage temperature: Freezer (–20 °C)