ContactUsLinkComponent
Fixed-angle titanium rotor package/assembly, MLA-55
Fixed-angle titanium rotor package/assembly, MLA-55
Supplier:  Beckman Coulter
Certificates
undefined
Fixed-angle titanium rotor package/assembly, MLA-55
Supplier:  Beckman Coulter
Specifications
  • Catalog No:
  • 521-4096
  • 521-4172
  • Capacity:
  • 108 ml
  • 108 ml
  • No. of tubes accepted:
  • 2/4/6/8×7 ml
  • 2/4/6/8×7 ml
  • Radius:
  • 84,5 mm
  • 84,5 mm
  • Max. RPM:
  • 55000
  • 55000