
SBAP (Succinimidyl 3-(bromoacetamido)propionate), Pierce™
Catalog # PIER22339
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Product Line:Pierce™
- Labeling Method:Chemical Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Haloacetyl, NHS Ester
- Spacer:Short
- Form:Solid
- Quantity:50 mg
- Type:Crosslinker
- Solubility:DMF, DMSO
Specification Test Results
About this item
:
:307.098 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:57159-62-3
Formula:C₉H₁₁BrN₂O₅
MDL Number:MFCD01863457